Orders LinkContactUsLinkComponent
VWR® Quantitative Grade Filter Paper Circles
VWR® Quantitative Grade Filter Paper Circles
VWR® Quantitative Grade Filter Paper Circles
Certificates
VWR® Quantitative Grade Filter Paper Circles
VWR® Quantitative Grade Filter Paper Circles
2 Options Available Below
Product Details & Documents
Quantitative filter papers are used for gravimetric analyses or filtrations where the precipitate is to be recovered.
  • Range of porosity available for filtering out precipitates
  • Flow rate varies between models
Sargent-Welch Quantitative
VWR® Grade #494
Porosity: Fine - Ashless
Flow Rate: Slow
Particle Retention: 1 μm
Applications: Useful for very fine precipitates. Ideal for BaSO₄, PbSO₄, SnS2, NiS, CaOH₂
VWR® Grade #474
Porosity: Medium - Ashless
Flow Rate: Medium
Particle Retention: 5 to 10 μm
Applications: For general quantitative analysis. Ideal for the filtration of AlPO₄, ZnNH₄PO₄, Ag₂CrO₄
Product Family Options
Product Information
  • Availability
    Price
  • 470201-456
    Quantitative Grade Filter Paper Circles, Sargent-Welch Quantitative, Diameter: 5.5 cm
    Quantitative Grade Filter Paper Circles, Sargent-Welch Quantitative, Diameter: 5.5 cm
    Log in for availability
    Log in for availability
    Paquete de 100
    Paquete de 100 - $546.87
    $546.87
    Paquete de 100
       $546.87
  • 470201-458
    Quantitative Grade Filter Paper Circles, Sargent-Welch Quantitative, Diameter: 7 cm
    Quantitative Grade Filter Paper Circles, Sargent-Welch Quantitative, Diameter: 7 cm
    Log in for availability
    Log in for availability
    Paquete de 100
    Paquete de 100 - $628.93
    $628.93
    Paquete de 100
       $628.93
  • 470204-444
    Quantitative Grade Filter Paper Circles, VWR® Grade #494, Diameter: 9 cm
    Quantitative Grade Filter Paper Circles, VWR® Grade #494, Diameter: 9 cm
    Discontinued
  • 470204-450
    Quantitative Grade Filter Paper Circles, VWR® Grade #474, Diameter: 12.5 cm
    Quantitative Grade Filter Paper Circles, VWR® Grade #474, Diameter: 12.5 cm
    Discontinued