ContactUsLinkComponent
PYREX® Standard Berzelius Tall Form Beakers
PYREX® Standard Berzelius Tall Form Beakers
Catálogo # 470211-360
Proveedor:  CORNING CA
Número de proveedor #: 1060-400
undefined
PYREX® Standard Berzelius Tall Form Beakers
Catálogo # 470211-360
Proveedor:  CORNING CA
Número de proveedor:  1060-400
Número de proveedor #: 1060-400
Iniciar sesión Para disponibilidad
Select an Option
400 ml (14 oz.)
Select an Option
Pyrex® Tall form Berzelius beakers with spout, graduated
Select an Option
470211-360
Select an Option
1060-400
Acerca de este artículo
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
Beaker with spout facilitates pouring. Graduated to indicate their approximate content.
Certificaciones: Meets ASTM E960 specifications.
Frequently Bought Together