Orders LinkContactUsLinkComponent
PYREX® Serological Pipettes, Plugged
Pipet, sterile serological Pyrex®, 2 ml, 0.2 ml, 0.01 ml, Individually Packed
  470211-592
 :   CORNING CA
 :  7077-2N
PYREX® Serological Pipettes, Plugged
  470211-592
PYREX® Serological Pipettes, Plugged
PYREX® Serological Pipettes, Plugged
Pipet, sterile serological Pyrex®, 2 ml, 0.2 ml, 0.01 ml, Individually Packed
$4,660.00
Paquete de 180 Case de 4 (180UN)
These sterile, glass pipettes, calibrated to deliver with blow-out; offer long slender tapered tips to make pipetting faster and easier.
Color-coded sizes.
Capacity Negative Grad. Divisions
WLS-69650-C 1.0 ml 0.2 ml 0.01 ml
WLS-69650-G 5.0 ml 1.0 ml 0.1 ml
WLS-69650-H 10.0 ml 2.0 ml 0.01 ml.
  • Capacity:
    2 ml
  • Description:
    Pipet, sterile serological Pyrex®
  • Packed:
    Individually Packed
  • Subdivisions:
    0.01 ml
  • Negative Graduations:
    0.2 ml