ContactUsLinkComponent
PYREX® Petri Culture Dishes
PYREX® Petri Culture Dishes
Catalog # 470211-460
Supplier:  CORNING CA
undefined
PYREX® Petri Culture Dishes
Catalog # 470211-460
Supplier:  CORNING CA
Supplier Number:  3160-60
Specifications
  • Descripción:
    Pyrex® Petri Culture Dishes, Dia. x H, mm: 60 x 15
  • Referencia:
    470211-460
  • Nº de proveedor:
    3160-60
Specifications
About this item
Pyrex® dishes have beaded edges for strength. The tops are marked with blue enamel, and the bottoms with white enamel for easy sorting. Bottoms also have an arrow reference point for serial dilutions or orientation. Dish is not airtight. Autoclavable (wet or dry). Complete Dish Assembly