PYREX® Funnels, with Short Stem, Glass, Corning
Supplier: CORNING CA
Certificates
About this item
PYREX®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°.
Frequently Bought Together