Orders LinkContactUsLinkComponent
PYREX® Watch Glasses
PYREX® Watch Glasses
PYREX® Watch Glasses
Supplier:  CORNING CA
Certificates
PYREX® Watch Glasses
PYREX® Watch Glasses
3 Options Available Below
Product Details & Documents
PYREX® borosilicate glass.
  • High chemical durability
  • Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
Packaging: One package has 12 watch glasses.
Documents
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • Watch glass
    65 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Log in for availability
    Each (12UN)
    Each (12UN) - $1,330.00
    $1,330.00
    Log in for availability
    Paquete de 12
    Paquete de 12 - $1,322.34
    $1,322.34
    Log in for availability
    Case de 12 (12UN)
    Case de 12 (12UN) - $1,435.27
    $1,435.27
    Each (12UN)
       $1,330.00
    Paquete de 12
       $1,322.34
    Case de 12 (12UN)
       $1,435.27
    Supplier Number: 9985-65
  • Watch glass
    75 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 12
    Paquete de 12 - $1,420.00
    $1,420.00
    Log in for availability
    Case de 12 (12UN)
    Case de 12 (12UN) - $1,524.31
    $1,524.31
    Paquete de 12
       $1,420.00
    Case de 12 (12UN)
       $1,524.31
    Supplier Number: 9985-75
  • Watch glass
    100 mm
    Discontinued
    Supplier Number: 9985-100
  • Watch glass
    125 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 12
    Paquete de 12 - $2,400.00
    $2,400.00
    Log in for availability
    Case de 6 (12UN)
    Case de 6 (12UN) - $2,581.69
    $2,581.69
    Paquete de 12
       $2,400.00
    Case de 6 (12UN)
       $2,581.69
    Supplier Number: 9985-125