Orders LinkContactUsLinkComponent
PYREX® Volumetric Flasks
PYREX® Volumetric Flasks
   Opciones sostenibles disponibles
Supplier:   CORNING CA
PYREX® Volumetric Flasks
Supplier:  CORNING CA
   Opciones sostenibles disponibles
Certificates
PYREX® Volumetric Flasks
6 Options Available Below
Product Details & Documents
PYREX® glass, clear, with snap-caps.
  • Heavy beads and tubing rings tooled for snap-cap fit
  • Graduation line is sharp and permanent
  • No snap caps for 1 and 2 ml sizes
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 470211-518
  • 470211-510
  • 470211-514
  • 470211-520
  • 470211-512
  • 470211-516
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Especificaciones
Product Family Options
Información del producto
  • Capacity
    Availability
    Price
  • 50 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $919.00
    $919.00
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $982.42
    $982.42
    Each (1UN)
       $919.00
    Case de 12 (1UN)
       $982.42
    Supplier Number: 5580-50
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-518
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 100 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $973.00
    $973.00
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,039.83
    $1,039.83
    Each (1UN)
       $973.00
    Case de 12 (1UN)
       $1,039.83
    Supplier Number: 5580-100
    Especificaciones:
    More
    Specifications
    Cat. No.
    470211-510
    Environmentally Preferable
    Environmental certification
  • 250 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,300.00
    $1,300.00
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,382.54
    $1,382.54
    Each (1UN)
       $1,300.00
    Case de 12 (1UN)
       $1,382.54
    Supplier Number: 5580-250
    Especificaciones:
    More
    Specifications
    Cat. No.
    470211-514
    Environmentally Preferable
    Environmental certification
  • 500 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,580.00
    $1,580.00
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,680.12
    $1,680.12
    Each (1UN)
       $1,580.00
    Case de 12 (1UN)
       $1,680.12
    Supplier Number: 5580-500
    Especificaciones:
    More
    Specifications
    Cat. No.
    470211-520
    Environmentally Preferable
    Environmental certification
  • 1000 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,850.00
    $1,850.00
    Log in Para disponibilidad
    Case de 6 (1UN)
    Case de 6 (1UN) - $1,976.55
    $1,976.55
    Each (1UN)
       $1,850.00
    Case de 6 (1UN)
       $1,976.55
    Supplier Number: 5580-1L
    Especificaciones:
    More
    Specifications
    Cat. No.
    470211-512
    Environmentally Preferable
    Environmental certification
  • 2000 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,020.00
    $3,020.00
    Log in Para disponibilidad
    Case de 4 (1UN)
    Case de 4 (1UN) - $3,223.20
    $3,223.20
    Each (1UN)
       $3,020.00
    Case de 4 (1UN)
       $3,223.20
    Supplier Number: 5580-2L
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-516
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact