Orders LinkContactUsLinkComponent
PYREX® Standard-Grade Beakers
PYREX® Standard-Grade Beakers
PYREX® Standard-Grade Beakers
Supplier:  CORNING CA
Certificates
PYREX® Standard-Grade Beakers
PYREX® Standard-Grade Beakers
6 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass.
  • PYREX®, Soda-lime glass, with spout
  • Very good chemical resistance
  • Good temperature resistance
Manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.
Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.
Certifications: Meet ASTM E960 specifications.
Ordering information: The variety pack includes one of each of the following beakers: 50, 100, 250, 600 and 1000 ml.
Product Family Options
Información del producto
  • Capacity
    Graduation Interval
    Graduation Range
    Availability
    Price
  • 10 ml (0.34 oz.)
    1 ml
    1 - 10 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $2,160.00
    $2,160.00
    Log in Para disponibilidad
    Case de 4 (12UN)
    Case de 4 (12UN) - $2,259.50
    $2,259.50
    Paquete de 12
       $2,160.00
    Case de 4 (12UN)
       $2,259.50
    Supplier Number: 1000-10
  • 20 ml (0.68 oz.)
    5 ml
    5 - 15 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $1,860.00
    $1,860.00
    Log in Para disponibilidad
    Case de 4 (12UN)
    Case de 4 (12UN) - $1,946.08
    $1,946.08
    Paquete de 12
       $1,860.00
    Case de 4 (12UN)
       $1,946.08
    Supplier Number: 1000-20
  • 30 ml
    10 ml
    5 - 25 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $1,810.00
    $1,810.00
    Log in Para disponibilidad
    Case de 4 (12UN)
    Case de 4 (12UN) - $1,926.20
    $1,926.20
    Paquete de 12
       $1,810.00
    Case de 4 (12UN)
       $1,926.20
    Supplier Number: 1000-30
  • 150 ml (5 oz.)
    10 ml
    20 - 140 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $1,730.00
    $1,730.00
    Log in Para disponibilidad
    Case de 4 (12UN)
    Case de 4 (12UN) - $1,849.44
    $1,849.44
    Paquete de 12
       $1,730.00
    Case de 4 (12UN)
       $1,849.44
    Supplier Number: 1000-150
  • 800 ml
    50 ml
    50 - 750 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 6
    Paquete de 6 - $1,890.00
    $1,890.00
    Log in Para disponibilidad
    Case de 4 (6UN)
    Case de 4 (6UN) - $2,011.69
    $2,011.69
    Paquete de 6
       $1,890.00
    Case de 4 (6UN)
       $2,011.69
    Supplier Number: 1000-800
  • 2000 ml
    100 ml
    200 - 1800 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $849.00
    $849.00
    Log in Para disponibilidad
    Case de 8 (1UN)
    Case de 8 (1UN) - $907.45
    $907.45
    Each (1UN)
       $849.00
    Case de 8 (1UN)
       $907.45
    Supplier Number: 1000-2L