Orders LinkContactUsLinkComponent
PYREX® Precision Burets
PYREX® Precision Burets
PYREX® Precision Burets
Supplier:  CORNING CA
Certificates
PYREX® Precision Burets
PYREX® Precision Burets
2 Options Available Below
Product Details & Documents
Suitable for Use Laboratories Not Requiring the Accuracy of Class A Models.
  • Dispensing Burets
  • Permanent graduation markings
  • Nonstick 2 mm Teflon stopcock ensures a smooth, leak-free operation
  • Dust cover included
Documents
Product Family Options
Información del producto
  • Capacity
    Graduation Interval
    Availability
    Price
  • 25 ml
    0.10 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $5,380.00
    $5,380.00
    Each (1UN)
       $5,380.00
    Supplier Number: 2122A-25
  • 50 ml
    0.10 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $5,450.00
    $5,450.00
    Each (1UN)
       $5,450.00
    Supplier Number: 2122A-50