Orders LinkContactUsLinkComponent
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Supplier:  CORNING CA
Certificates
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
3 Options Available Below
Product Details & Documents
Manufactured from borosilicate glass.
Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).
Documents
Product Family Options
Información del producto
  • Capacity
    Stopper No.
    Availability
    Price
  • 50 ml
    1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $784.00
    $784.00
    Each (1UN)
       $784.00
    Supplier Number: 4060-50
  • 250 ml
    5
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $524.00
    $524.00
    Log in Para disponibilidad
    Case de 24 (1UN)
    Case de 24 (1UN) - $555.96
    $555.96
    Each (1UN)
       $524.00
    Case de 24 (1UN)
       $555.96
    Supplier Number: 4060-250
  • 500 ml
    6
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (6UN)
    Each (6UN) - $585.00
    $585.00
    Log in Para disponibilidad
    Case de 24 (6UN)
    Case de 24 (6UN) - $654.36
    $654.36
    Each (6UN)
       $585.00
    Case de 24 (6UN)
       $654.36
    Supplier Number: 4060-500