Ward's® Pulleys Activity Kit: Exploring Force, Mass and Simple Machines
Catálogo # 470013-290
Proveedor: Avantor
Especificaciones
- Descripción:Ward's® Pulleys Activity Kit: Exploring Force, Mass and Simple Machines
- Referencia:470013-290
- Nº de proveedor:WARD470013-290
Especificaciones
Acerca de este artículo
The activity includes one triple sheave, one double sheave, and two single sheave pulleys, as well as a beautifully illustrated activity guide with questions, making it ideal for single student study or as a demonstration unit.
- Students assemble and observe how pulleys can make work easier
Pulleys were among the first machines invented by man. They used them in ingenious ways to help solve problems and perform tasks more easily. In this economical activity, students construct both single and compound pulley systems and observe their function. Students explore the concepts of gravity, force, friction, mechanical advantage, and more.