Pulleys
Supplier: VWR International
Jump to:
About this item
These free-running plastic pulleys are durable in design.
- Plastic Construction
- Durable
- Nickel-Plated Steel Blocks with Hooks
The sheaves are 50 mm in Ø and made of rigid plastic.
The nickel-plated steel blocks have hooks at the top and bottom.
4 Options Available Below
Product Details & Documents
These free-running plastic pulleys are durable in design.
- Plastic Construction
- Durable
- Nickel-Plated Steel Blocks with Hooks
The sheaves are 50 mm in Ø and made of rigid plastic.
The nickel-plated steel blocks have hooks at the top and bottom.
Product Family Options
Product Information
- AvailabilityPrice
- 470235-738Pulley, Single SheavePulley, Single SheaveEach (1UN)$36.86Paquete de 20 (1UN)$40.18
- 470104-442Pulley, Double Sheave
- 470235-736Pulley, Triple SheavePulley, Triple SheaveEach (1UN)$87.24Paquete de 15 (1UN)$95.10
- 470005-698Pulley, Quadruple Sheave