Pteridium, Rhizome Slide
Pteridium, Rhizome, Cross section, Quadruple
470181-176
: Avantor
Fern Species Common in Temperate and Subtropical Areas
- Identifying features clearly distinguishable
- Rigorous quality control standards
Bracken. Showing a dictyostele.
- Description:Pteridium, Rhizome
- Stain:Quadruple
- Orientation:Cross section