Pteridium, Mature Leaflet Slide
Pteridium, Mature Leaflet, Cross section, Quadruple
470181-180
: Avantor
Fern Species Common in Temperate and Subtropical Areas
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows fully developed sporangia.
- Description:Pteridium, Mature Leaflet
- Stain:Quadruple
- Orientation:Cross section