Prokaryote Survey Slide Set
Prokaryote Survey Slide Set, 12, Monera
Catalog # 470177-698
Supplier: Avantor
Jump to:
Product Details & Documents
An Ideal Guide to the Major Prokaryote Groups
- Identifying features clearly distinguishable
- Rigorous quality control standards
All are slides are prepared as smears unless otherwise specified.
Ordering information: Set includes Archaebacteria: Mixed forms; Eubacteria: Typical bacillus, Typical gram + cocci, Typical gram – cocci, Typical spirillum, Cocci in clumps (Staphylococci), Cocci in chains (Streptococci), Bacillus megaterium, Micrococcus luteus, Treponema sp., Root nodule-legume bacteria (sect); Cyanobacteria: Anabaena (wm), and Anacystis (wm).
Especificaciones
- Type:Monera
- Description:Prokaryote Survey Slide Set
- No. of Slides:12