Orders LinkContactUsLinkComponent
Probe And Seeker
Probe And Seeker
Probe And Seeker
Certificates
Probe And Seeker
Probe And Seeker
2 Options Available Below
Product Details & Documents
This tool performs double duty, with a curved probe at one end and a flat spade at the other to gently separate muscles without tearing or cutting. Stainless steel.
Product Family Options
Product Information
  • Material
    Length
    Tip Style
    Availability
    Price
  • 470018-924
    Probe And Seeker
    Probe And Seeker
    Stainless steel
    152 mm (6")
    Bent
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $43.71
    $43.71
    Each (1UN)
       $43.71
    Supplier Number: 140958
  • 470122-006
    Probe And Seeker
    Probe And Seeker
    Stainless steel
    165 mm (6.5")
    Curved
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $55.44
    $55.44
    Each (1UN)
       $55.44
    Supplier Number: WLS26907