Equilateral Acrylic Prisms
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
Acerca de este artículo
Demonstrate refraction and internal reflection.
- Highly polished sides
- High optical clarity
Selection of equilaterial and right angle acrylic prisms.
4 Options Available Below
Product Details & Documents
Demonstrate refraction and internal reflection.
- Highly polished sides
- High optical clarity
Selection of equilaterial and right angle acrylic prisms.
Product Family Options
Información del producto
- AvailabilityPrice
- 470004-204Prism, Equilateral/Acrylic, 25 mm x 25 mmSupplier Number: PAE025
- 470013-002Prism, Equilateral/Acrylic, 75 mm x 25 mmSupplier Number: PAE075
- 470101-250Prism, Equilateral/Acrylic, 100 mm x 25 mmSupplier Number: PAE100
- 470101-252Prism, Equilateral/Acrylic, 150 mm x 25 mmSupplier Number: PAE150