ContactUsLinkComponent
Principles and Practice of Agarose Gel Electrophoresis Kit
Principles and Practice of Agarose Gel Electrophoresis Kit
Catalog # 470121-714
Supplier:  EDVOTEK, INC.
undefined
Principles and Practice of Agarose Gel Electrophoresis Kit
Catalog # 470121-714
Supplier:  EDVOTEK, INC.
Supplier Number:  101
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Descripción:
    Principles and Practice of Agarose Gel Electrophoresis Kit
  • No. of Lab Groups:
    8
  • Referencia:
    470121-714
  • Nº de proveedor:
    101
Specifications
About this item
The Principles and Practice of Agarose Gel Electrophoresis Kit demonstrates how electrophoresis separates molecules on the basis of size and charge.
  • Safe, Colorful, and Fast
  • Time Required: 45 Minutes
  • Kit Demonstrates Electrophoresis
  • Instructions Included
  • Ready-To-Load Dye Samples Included
It's a safe, colorful, fast, and simple way to teach a technique that engages students.
The activity requires approximately 45 minutes.
Ordering information: This kit includes instructions, Ready-to-Load dye samples, agarose powder, practice gel-loading solution, electrophoresis buffer, and microtipped transfer pipettes. An electrophoresis apparatus, power supply, microwave, or hot plate is required.
Delivery: Once you receive this package, store Ready-to-Load QuickStrip samples in the refrigerator.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator