AC/DC power supply
AC/DC power supply, 0 – 6 V DC
1, 2, 3, 4, 5, 6 V AC, 5A , 110/220 V AC
1, 2, 3, 4, 5, 6 V AC, 5A , 110/220 V AC
Catalog # 470125-556
Supplier: VWR International
Jump to:
Product Details & Documents
Ideal for charging wet cell batteries, electroplating, electrolysis, and many other applications.
Output is electronically protected and the line fuse protects against internal shorts. A front panel voltage knob makes adjustments easy and color-coded 4mm banana jacks provide output. Includes a three-wire grounded cord and plug.
Certifications: CSA Approved
Especificaciones
- Voltage:0 – 6 V DC
1, 2, 3, 4, 5, 6 V AC - Description:AC/DC power supply
- Voltage Input:110/220 V AC
- Current:5A