Polypropylene Powder Funnels
Powder Funnel, Polypropylene, 80 mm diameter, Scienceware
Supplier: VWR International
Jump to:
Product Details & Documents
These unbreakable polypropylene powder funnels are ideal for student use. External ribs prevent air lock. Can be autoclaved. Stem length is 35 mm.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Description:Powder Funnel, Polypropylene, 80 mm diameter, Scienceware