Porous Cups
Supplier: VWR International
Jump to:
About this item
Porous cylindrical cups.
- Made of ceramic
Pore diameter is approximately 0.5 µm.
3 Options Available Below
Product Details & Documents
Porous cylindrical cups.
- Made of ceramic
Pore diameter is approximately 0.5 µm.
Product Family Options
Product Information
- Diameter ×HAvailabilityPrice
- 470104-080Porous cups
- 470148-830Porous cupsPorous cups40×90 mmEach (1UN)$78.48Case de 12 (1UN)$85.54
- 470149-184Porous pots, cylindricalPorous pots, cylindrical25×76 mmEach (1UN)$60.19Case de 24 (1UN)$65.61