ContactUsLinkComponent
Porcelain Casseroles
  470148-900
undefined
Porcelain Casseroles
  470148-900
 :  J-269
  • Descripción:
    Porcelain Casserole, Size 2, 60 mL
  • Referencia:
    470148-900
  • Nº de proveedor:
    J-269
High-Quality Labware, Bargain Prices
Our laboratory porcelain is high-quality, yet affordably priced. This casserole has a lip, pouring spout and flat porcelain handle. It is glazed entirely on the inside and outside with the exception of the rim. This piece can withstand relatively high temperatures in standard lab settings. Note: Fragile, use care when handling.