Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Especificaciones
- Descripción:Reagent Refill
- No. of Students:50
- Referencia:470003-958
- Nº de proveedor:R-5418
Especificaciones
Acerca de este artículo
Study water quality of lakes, streams, ponds, and even aquariums.
- Test for ammonia, dissolved oxygen, nitrate, and pH
- Individually sealed foil reagent tablets
- Materials for 50 students
With this activity, students will investigate the natural process and variables that create changes in water quality over a period of time. They will collect samples from any local freshwater source, then use reagent tablets in individually sealed foil strips to test the ammonia, dissolved oxygen, and nitrate contents, as well as the pH of the sample. Materials for 50 students.