Polystyrene Spot Plate
Polystyrene spot plate
Catalog # 470002-082
Supplier: Avantor
Jump to:
Product Details & Documents
Chemically inert and unbreakable.
- Can be used with concentrated acids
Each plate has 12 ¹/₃" diameter depressions. Size: 4¹/₂×3¹/₂"
Packaging: Package of 12.
Especificaciones
- Description:Polystyrene spot plate