Orders LinkContactUsLinkComponent
Polypropylene Volumetric Flasks with Screw Caps
Polypropylene Volumetric Flasks with Screw Caps
Certificates
Polypropylene Volumetric Flasks with Screw Caps
Polypropylene Volumetric Flasks with Screw Caps
2 Options Available Below
Product Details & Documents
Excellent Chemical Resistance
  • Calibrated “To Contain” and “To Deliver” at 20°C (68°F)
  • No meniscus to confuse readings
  • Threaded neck with polypropylene screw closure
Autoclavable (Remove closure before autoclaving).
 : Cap included but not shown.
Product Family Options
Información del producto
  • Capacity
    Availability
    Price
  • 50 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,220.00
    $1,220.00
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,435.27
    $1,435.27
    Each (1UN)
       $1,220.00
    Case de 12 (1UN)
       $1,435.27
    Supplier Number: 4000-0050
  • 500 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,950.00
    $1,950.00
    Log in Para disponibilidad
    Case de 8 (1UN)
    Case de 8 (1UN) - $2,295.81
    $2,295.81
    Each (1UN)
       $1,950.00
    Case de 8 (1UN)
       $2,295.81
    Supplier Number: 4000-0500