Polypropylene Separatory Funnels
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
Acerca de este artículo
Resists Virtually All Chemicals
- Stem length 45 mm
- Includes a leak-proof threaded screw cap
- Not graduated
The PTFE plug and housing eliminate the need for lubrication and thus reduce the possibility of contamination. The phase interface is visible down to the stopcock. Screw closures are leak-proof, and unbreakable. Autoclavable except for stopcock assembly, which is easily removed.
3 Options Available Below
Product Details & Documents
Resists Virtually All Chemicals
- Stem length 45 mm
- Includes a leak-proof threaded screw cap
- Not graduated
The PTFE plug and housing eliminate the need for lubrication and thus reduce the possibility of contamination. The phase interface is visible down to the stopcock. Screw closures are leak-proof, and unbreakable. Autoclavable except for stopcock assembly, which is easily removed.
Product Family Options
Información del producto
- CapacityStopcock SizeAvailabilityPrice
- 100 ml3 mmEach (1UN)$630.96Case de 20 (1UN)$687.74
- 500 ml4 mmEach (1UN)$368.16Case de 12 (1UN)$401.29
- 500 ml4 mmEach (1UN)$368.16Case de 12 (1UN)$401.29