Polypropylene Scoops
Polypropylene Scoops, Scoop (L x W x D): 8-1/2 x 5 x 3", Handle Length: 4"
Supplier: BEL-ART PRODUCTS SE
Número de proveedor #: F36756-0000_SE
Jump to:
Product Details & Documents
Molded of strong, smooth polypropylene.
Steam autoclavable at 120°C. Scoop size: 8 1/2" x 5" x 3". Handle Length: 4".
Delivery information: Package of six.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Description:Polypropylene Scoops, Scoop (L x W x D): 8-1/2 x 5 x 3", Handle Length: 4"