Polypropylene Petri Dish Rack
Polypropylene Petri Dish Rack
Catalog # 470020-886
Supplier: BEL-ART PRODUCTS SE
Número de proveedor #: F18991-0000_SE
Jump to:
Product Details & Documents
This autoclavable polypropylene rack holds forty-two 100 mm dishes. It has a small footprint, is stackable to conserve counter space, and it features a handle for easy transport. Size: 13"L x 8 1/4"W x 7"H.
Especificaciones
- Description:Polypropylene Petri Dish Rack