Orders LinkContactUsLinkComponent
Polypropylene Cylinders with Handle
Polypropylene Cylinders with Handle
Polypropylene Cylinders with Handle
Certificates
Polypropylene Cylinders with Handle
2 Options Available Below
Product Details & Documents
Handle Provides Safe, Solid Grip and Superior Control When Pouring.
  • Double scale
  • Octagonal base
  • Autoclavable up to 121°C
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 470121-904
  • 470121-906
  • Form:
  • Tall
  • Tall
Especificaciones
Product Family Options
Información del producto
  • Capacity
    Graduation Interval
    Availability
    Price
  • 470121-904
    Polypropylene Cylinders with Handle
    Polypropylene Cylinders with Handle
    1000 ml
    10 mL
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $926.00
    $926.00
    Each (1UN)
       $926.00
    Supplier Number: F28461-1000_SE
    Especificaciones:
    More
    Specifications
    Cat. No.
    470121-904
    Form
    Tall
  • 470121-906
    Polypropylene Cylinders with Handle
    Polypropylene Cylinders with Handle
    2000 ml
    20 mL
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,810.00
    $1,810.00
    Each (1UN)
       $1,810.00
    Supplier Number: F28461-2000_SE
    Especificaciones:
    More
    Specifications
    Cat. No.
    470121-906
    Form
    Tall