Polypropylene Buchner Funnel
Polypropylene Buchner Funnel
Catalog # 470174-670
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: 57105-PK/12
Jump to:
Product Details & Documents
Useable With or Without Filter Paper.
- Uses 7.0 cm diameter filter paper
Two-piece HDPE funnel with sealed-in, polyethylene filter plate of coarse porosity offers good retention and fast flow under vacuum.
Specifications
- Description:Polypropylene Buchner Funnel