Medegen Tri-Pour Beakers
470146-288
- Intervalo de graduación:50 ml
- Rango de graduación:50 - 1000 ml
- Capacidad:1000 ml
- Referencia:470146-288
- Nº de proveedor:25384-160
Shatterproof Polypropylene.
- Chemically resistant to commonly used solvents and reagents
- Clearly marked graduations in both mL and oz.
- Autoclavable
These unique beakers feature three indented pouring spouts for more accurate dispensing of fluids with less dripping. They also feature flat bottoms for magnetic stirring.