Polygrid Petri Dish Carrying Rack
Catálogo # 470105-610
Proveedor: BEL-ART PRODUCTS SE
Especificaciones
- Descripción:Petri Dish Carrying Rack, Poxygrid Holds 20 Dishes
- Referencia:470105-610
- Nº de proveedor:H189780000
Especificaciones
Acerca de este artículo
This epoxy coated steel wire rack holds 20 plastic petri dishes. Petri dishes are easily loaded and emptied from the top. The wide front opening permits easy access but prevents dishes from sliding out. Not suitable for glass petri dishes.