Polyethylene Test Tube Supports with Draining Pins
Proveedor: PRECISION MOULDED POLYMERS LTD
Certificados
Acerca de este artículo
These supports keep test tubes secure.
- Supports Hold up to 12 Test Tubes
- Five vertical drying pins
- Supports Keep Test Tubes Secure
They hold up to 12 test tubes.
They have five vertical drying pins.
Frequently Bought Together