Polypropylene Funnels, Dynalon
Funnel, Poly, 114 mL (4 oz)
Catalog # 470161-302
Supplier: DYNALON
Número de proveedor #: 412334-0002
Jump to:
Product Details & Documents
These Polypropylene Funnels are chemical and acid resistant and can be sterilized. These funnels are offered in four different diameters/capacities to meet all your funneling needs.
Especificaciones
- Description:Funnel, Poly, 114 mL (4 oz)