Polyethylene Dropping Bottles
59 ml (2 oz.), LDPE
Catalog # 470147-500
Supplier: DYNALON
Supplier #: 605065-0020
Jump to:
Product Details & Documents
Economy LDPE dropper bottles.
- Translucent LDPE dropper bottles
- Sturdy white dropping cap
- Flip up spout
Dropper bottles with hinged dispenser caps for dispensing indicator solutions, dyes, and more.
Specifications
- Capacity:59 ml (2 oz.)
- Material:LDPE