Polyethylene Disposable Apron
Polyethylene Apron, Clear, 28×46"
470156-174
Supplier #: DAP125
One-piece, chemical-resistant aprons can be worn once and thrown away.
- Disposable
- Convenient Dispenser Box
- One-Piece
- Chemical-Resistant
Keep fresh aprons handy at all times with a convenient dispenser box that issues aprons one at a time.
The one-piece apron design eliminates folds, hems, and seams that can trap bacteria or dirt.
: One box contains 100 aprons.
- Color:Clear
- Description:Polyethylene Apron
- W×L:28×46"