Orders LinkContactUsLinkComponent
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Low-density polyethylene bottles and spouts with sealer caps that snap on securely. Spout openings are standard 0.030" and can be enlarged with scissors.
Product Family Options
  • Description
  • Bottle, Dispensing, 30 mL
    Paquete de 12
    Paquete de 12 - $256.91
    $256.91
    Paquete de 12
       $256.91
     : 300 & 351
  • Bottle, Dispensing, 59 mL
    Paquete de 12
    Paquete de 12 - $300.08
    $300.08
    Paquete de 12
       $300.08
     : 301 & 351
  • Bottle, Dispensing, 118 mL
    Paquete de 12
    Paquete de 12 - $368.83
    $368.83
    Paquete de 12
       $368.83
     : 395 & 93
  •    
    Paquete de 12
    Paquete de 12 - $28.65
    $28.65
    Case de 6 (12UN)
    Case de 6 (12UN) - $171.90
    $171.90
    Paquete de 12
       $28.65
    Case de 6 (12UN)
       $171.90
     : F11637-0008_SE