Plumb Bob
Plumb Bob
Catalog # 470123-240
Supplier: STEINER ENTERPRISES SE
Número de proveedor #: S70200
Jump to:
Product Details & Documents
Nickel-plated steel. With center cavity type screw cap.
- Length: 10 cm
- Diameter: 2 cm
- Weight: 227 g
Especificaciones
- Description:Plumb Bob