Plastic Vernier Caliper, Walter Stern
470205-490
- Descripción:Plastic Vernier Caliper
- Referencia:470205-490
- Nº de proveedor:602C
Full-Size Vernier Caliper, durable plastic with metal parts.
Perfect for student use.
English and metric scales read to 5 inches in 1/16 " increments or 12cm in 1mm increments. Vernier permits measurements to 1/10 of a division.