Plastic Slide Box for 100 Slides
470150-636
- Capacidad:100
- Descripción:Plastic Slide Box, 100 Slides
- Referencia:470150-636
- Nº de proveedor:HS15994A
Excellent For Storing Standard Sized Slides
- Keep slides debris-free
- Store slides neatly and efficiently
Store 100 standard slides in this two-piece polystyrene case with steel pin hinges and a steel latch. The bottom is cork lined to protect slides. The slots are numbered and correspond with an index list on lid.