Plastic Microscope Slides
Plastic slides, 0.5 mm, 3×1"
Catalog # 470014-584
Supplier: PRECISION MOULDED POLYMERS LTD
Número de proveedor #: AM9330.00000.00000
Jump to:
Product Details & Documents
Standard Sized Slides with Less Injury Potential
- Quality slide making supplies
- Limit injuries in the lab
Safer than glass, these optically clear plastic slides will not break, scratch, or cut fingers. Package of 72 slides.
Especificaciones
- Description:Plastic slides
- L×W:3×1"
- Thickness:0.5 mm