Ward's® Plastic Labeledge Trays
: Avantor
Ideal for storage or display of larger specimens
- Multiple sizes available
- Display and storage purposes
Tough white polystyrene trays are available in three sizes to hold various samples. Trays feature slanted fronts for labels.
Ideal for storage or display of larger specimens
- Multiple sizes available
- Display and storage purposes
Tough white polystyrene trays are available in three sizes to hold various samples. Trays feature slanted fronts for labels.
Product Family Options
- DescriptionWidthHeightLength
- Small labeledge tray2½"⅝"2½"-: B19180/1-SP
- Large labeledge tray4½"¾"4½"Each (1UN)$19.05Paquete de 100 (1UN)$1,905.00: -