Plastic Beads
Supplier: Avantor
Jump to:
Acerca de este artículo
Packages of approximately 100.
- 4 mm Red bead = Acrylic
- 8 mm Navy bead = Acrylic
- 12 mm White bead = Acetate
- Autoclavable
3 Options Available Below
Product Details & Documents
Packages of approximately 100.
- 4 mm Red bead = Acrylic
- 8 mm Navy bead = Acrylic
- 12 mm White bead = Acetate
- Autoclavable
Product Family Options
Información del producto
- ØAvailabilityPrice
- 470015-258Plastic Beads
- 470023-956Plastic Beads
- 470023-958Plastic Beads