ContactUsLinkComponent
Plastic 473 ml Wide Mouth Jar
Catalog # 470189-441
Supplier:  3G PACKAGING, INC.
Plastic 473 ml Wide Mouth Jar
Catalog # 470189-441
Supplier:  3G PACKAGING, INC.
Supplier Number:  6986400
Specifications
  • Descripción:
    Plastic 473 mL Wide Mouth Jar
  • Referencia:
    470189-441
  • Nº de proveedor:
    6986400
Specifications
About this item
This sturdy plastic 473 mL (16 oz.) jar has absolutely straight sides, making it ideal for storing bulky materials.
Ordering information: Package of 24, Caps are not included.