Plant Leaf Impression
Plant Leaf Impression, 3"
Catalog # 470225-094
Supplier: Avantor
Jump to:
Product Details & Documents
Detailed angiosperm leaf impression.
- Eocene
Display quality, large angiosperm leaf impressions; assorted varieties. Average size: 3" on 4×5"
Especificaciones
- Description:Plant Leaf Impression
- Specimen Dimensions:3"