Ward's® Plant And Animal Cell Structure Poster Sets
470023-242
: Avantor
- Anchura:53 cm
- Descripción:Cell Structure Set
- Longitud:86 cm
- Referencia:470023-242
- Se ajuste:Unlaminated
- Nº de proveedor:WARD470023-242
Large Posters Display Intricate Features Of Cell In Vivid Color
- Bright, easy-to-see illustrations
The illustrations of a typical plant cell or a typical animal cell have clearly labeled parts. The two charts also feature descriptions of the structure and function of each organelle and its relation to the entire cell structure. The plant cell poster shows the cell wall, large vacuole, chloroplasts, nucleus, ribosomes, Golgi complex, mitochondria, and more; the animal cell poster shows the plasma membrane, nucleus, ribosomes, Golgi complex, mitochondria, centrioles, cytoskeleton, and more.