LaMotte® Plankton Net
Plankton Net, 13Ø×38D cm
Catalog # 470203-064
Supplier: LA MOTTE CHEMICAL PROD CO SE
Número de proveedor #: 1063
Jump to:
Product Details & Documents
Use this cone-shaped fine-mesh nylon net to let water pass while you filter out plankton.
- 10 mesh, 153 micron nylon cloth
- Clear, conical tube attached at end of the net allows plankon viewing
- Net mouth braced by sturdy stainless steel ring and harness
A brass ring and rope harness keep the throat of the net open while the net is being towed. A conical, graduated tube (50 ml capacity) is secured to the end of the net for sample collection.
Delivery information: Includes spare graduated tube.
Especificaciones
- Description:Plankton Net
- Size:13Ø×38D cm