Plankton Dip Net
Plankton Dip Net, Handle Length 51 cm, 19 cm, 7 cm
Catalog # 470106-844
Supplier: ED CUMINGS, INC.
Número de proveedor #: BW-20-X
Jump to:
Product Details & Documents
Small Mesh Size to Collect More Specimens
- Reinforced for More Durability
Collect microscopic specimens with this extra-fine mesh dip net. The net is re-inforced with a vinyl net saver, and a wooden 51 cm handle provides an excellent grip. Size: 19×15×7 cm.
Especificaciones
- Width:7 cm
- Description:Plankton Dip Net, Handle Length 51 cm
- Length:19 cm