Graduated Transfer Pipettes
Supplier: C & A SCIENTIFIC CO INC
Jump to:
Acerca de este artículo
Designed for consisent drop sizes.
- Economical and convenient polyethylene
- Inert to most biologicals and acids
- Non-sterile
3 Options Available Below
Product Details & Documents
Designed for consisent drop sizes.
- Economical and convenient polyethylene
- Inert to most biologicals and acids
- Non-sterile
Product Family Options
Información del producto
- CapacityStem DiameterTotal lengthDrops per mLGraduationsBulb DrawAvailabilityPrice
- 470017-122Pipette graduated, 182971Pipette graduated, 1829715.0 ml5 mm150 mm250.25 ml3.1 mlLog in Para disponibilidadPaquete de 500$482.91Supplier Number: PTP-02
- 470020-860Pipette graduated, 182977Pipette graduated, 1829775.0 ml5 mm150 mm250.25 ml3.1 mlLog in Para disponibilidadPaquete de 100$159.20Supplier Number: -
- 470152-946Pipette dropper graduated, 2988402Pipette dropper graduated, 29884025.0 ml5 mm150 mm250.25 ml3.1 mlLog in Para disponibilidadPaquete de 1$41.83Supplier Number: -