Pipette Pumps
Supplier: VWR International
Jump to:
Acerca de este artículo
Safe and accurate.
- Easily disassembled for cleaning
- Color-coded for pipette size selection
- Resist acid and alkalis
These pumps are easy to use in one-handed operation. The thumbwheel is simply rotated for precise filling or dispensing. Measuring pipettes fit smoothly into tapered chuck and a plunger may be used for quick dispensation.
3 Options Available Below
Product Details & Documents
Safe and accurate.
- Easily disassembled for cleaning
- Color-coded for pipette size selection
- Resist acid and alkalis
These pumps are easy to use in one-handed operation. The thumbwheel is simply rotated for precise filling or dispensing. Measuring pipettes fit smoothly into tapered chuck and a plunger may be used for quick dispensation.
Product Family Options
Información del producto
- AvailabilityPrice
- 470145-2981 & 2 ml Pipette Pump (Blue)1 & 2 ml Pipette Pump (Blue)Each (1UN)$834.00Case de 12 (1UN)$1,181.90
- 470145-2965 & 10 ml Pipette Pump (Green)5 & 10 ml Pipette Pump (Green)Each (1UN)$894.00Case de 12 (1UN)$1,266.97
- 470145-29425 ml Pipette Pump (Red)25 ml Pipette Pump (Red)Each (1UN)$1,140.00Case de 12 (1UN)$872.99